• USA Home
  • 416568 - Bisphenol A propoxylate/ethoxylate

EMAIL THIS PAGE TO A FRIEND
416568 Sigma-Aldrich

Bisphenol A propoxylate/ethoxylate

average Mn ~548, EO/phenol 1, PO/phenol 2

Purchase

Properties

mol wt   average Mn ~548
composition   EO/phenol, 1
  PO/phenol, 2
refractive index   n20/D 1.523 (lit.)
density   1.075 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?