• USA Home
  • 416649 - Samarium(III) bromide hexahydrate

EMAIL THIS PAGE TO A FRIEND
416649 Sigma-Aldrich

Samarium(III) bromide hexahydrate

99.9%

Purchase

Properties

assay   99.9%
SMILES string   O.O.O.O.O.O.Br[Sm](Br)Br
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?