• USA Home
  • 417157 - Allyl(diisopropylamino)dimethylsilane

EMAIL THIS PAGE TO A FRIEND
417157 Sigma-Aldrich

Allyl(diisopropylamino)dimethylsilane

98%

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Other Organosilicon
InChI Key   GSTVYECJWYQBKF-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.447 (lit.)
bp   79-81 °C/13 mmHg (lit.)
density   0.815 g/mL at 25 °C (lit.)

Description

Packaging

Packaged in glass bottles

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
144 °F
Flash Point(C) 
62 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?