EMAIL THIS PAGE TO A FRIEND
417629 Sigma-Aldrich

N,N′-Dipropylurea

97%

  • CAS Number 623-95-0

  • Linear Formula CH3CH2CH2NHCONHCH2CH2CH3

  • Molecular Weight 144.21

  •  Beilstein/REAXYS Number 1751900

  •  EC Number 210-821-8

  •  MDL number MFCD00192114

  •  PubChem Substance ID 24866166

Purchase

Properties

assay   97%
mp   104-106 °C (lit.)
SMILES string   CCCNC(=O)NCCC
InChI   1S/C7H16N2O/c1-3-5-8-7(10)9-6-4-2/h3-6H2,1-2H3,(H2,8,9,10)
InChI key   AWHORBWDEKTQAX-UHFFFAOYSA-N
Gene Information   human ... EPHX2(2053)
mouse ... Ephx2(13850)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?