• USA Home
  • 418234 - Zinc 2,9,16,23-tetrakis(phenylthio)-29H,31H-phthalocyanine

EMAIL THIS PAGE TO A FRIEND
418234 Sigma-Aldrich

Zinc 2,9,16,23-tetrakis(phenylthio)-29H,31H-phthalocyanine

Dye content 85 %

  • CAS Number 77447-43-9

  • Empirical Formula (Hill Notation) C56H32N8S4Zn

  • Molecular Weight 1010.56

  •  MDL number MFCD00192580

  •  PubChem Substance ID 24866213

Purchase

Properties

composition   Dye content, 85%
mp   270 °C (dec.) (lit.)
absorption   λmax 692 nm
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?