• USA Home
  • 418358 - 4-Nonene (cis+trans)

EMAIL THIS PAGE TO A FRIEND
418358 Sigma-Aldrich

4-Nonene (cis+trans)

95%

  • CAS Number 2198-23-4

  • Linear Formula CH3(CH2)3CH=CHCH2CH2CH3

  • Molecular Weight 126.24

  •  Beilstein/REAXYS Number 1733773

  •  EC Number 218-595-2

  •  MDL number MFCD00009507

  •  PubChem Substance ID 24866220

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   KPADFPAILITQBG-VQHVLOKHSA-N
assay   95%
refractive index   n20/D 1.419 (lit.)
bp   144-146 °C(lit.)
density   0.732 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3295C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
86 °F
Flash Point(C) 
30 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?