EMAIL THIS PAGE TO A FRIEND
419788 Sigma-Aldrich

7-Methoxyflavanone

98%

Purchase

Properties

Related Categories Benzopyrans, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
InChI Key   VYESEQLQFXUROZ-UHFFFAOYSA-N
assay   98%
mp   89-91 °C (lit.)
Gene Information   human ... CYP19A1(1588)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?