• USA Home
  • 41984 - Fmoc-3-(2-quinolyl)-DL-Ala-OH

EMAIL THIS PAGE TO A FRIEND
41984 Sigma-Aldrich

Fmoc-3-(2-quinolyl)-DL-Ala-OH

≥96.0% (HPLC)

Synonym: Fmoc-3-(2-quinolyl)-DL-alanine

  • CAS Number 401514-70-3

  • Empirical Formula (Hill Notation) C27H22N2O4

  • Molecular Weight 438.47

  •  MDL number MFCD03095516

  •  PubChem Substance ID 57650092

  •  NACRES NA.26

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
Quality Level   100
assay   ≥96.0% (HPLC)
form   powder
application(s)   peptide synthesis: suitable
color   white to faintly yellow
functional group   Fmoc
storage temp.   2-8°C
SMILES string   OC(=O)C(Cc1ccc2ccccc2n1)NC(=O)OCC3c4ccccc4-c5ccccc35
InChI   1S/C27H22N2O4/c30-26(31)25(15-18-14-13-17-7-1-6-12-24(17)28-18)29-27(32)33-16-23-21-10-4-2-8-19(21)20-9-3-5-11-22(20)23/h1-14,23,25H,15-16H2,(H,29,32)(H,30,31)
InChI key   IDOMHXAHHXHYMO-UHFFFAOYSA-N

Description

Packaging

250 mg in poly bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?