• USA Home
  • 419877 - Dimethylditetradecylammonium bromide

EMAIL THIS PAGE TO A FRIEND
419877 Sigma-Aldrich

Dimethylditetradecylammonium bromide

97%

Synonym: Ditetradecyldimethylammonium bromide

  • CAS Number 68105-02-2

  • Linear Formula [CH3(CH2)13]2N(Br)(CH3)2

  • Molecular Weight 518.74

  •  Beilstein/REAXYS Number 4050300

  •  EC Number 268-474-3

  •  MDL number MFCD00191429

Purchase

Properties

InChI Key   IRMGVPILCPGYNQ-UHFFFAOYSA-M
assay   97%
mp   167-169 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

40225 Dimethylditetradecylammonium bromide, ≥97.0% (NT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?