• USA Home
  • 420077 - Butyl (S)-(−)-lactate

EMAIL THIS PAGE TO A FRIEND
420077 Sigma-Aldrich

Butyl (S)-(−)-lactate

98%

Synonym: (−)-Butyl L-lactate

  • CAS Number 34451-19-9

  • Linear Formula CH3CH(OH)CO2(CH2)3CH3

  • Molecular Weight 146.18

  •  Beilstein/REAXYS Number 1721597

  •  EC Number 252-036-3

  •  MDL number MFCD00066454

  •  PubChem Substance ID 24866308

Purchase

Properties

vapor density   4 (20 °C, vs air)
assay   98%
optical activity   [α]20/D −12°, neat
autoignition temp.   720 °F
bp   185-187 °C (lit.)
mp   −28 °C (lit.)
density   0.984 g/mL at 25 °C (lit.)
SMILES string   CCCCOC(=O)[C@H](C)O
InChI   1S/C7H14O3/c1-3-4-5-10-7(9)6(2)8/h6,8H,3-5H2,1-2H3/t6-/m0/s1
InChI key   MRABAEUHTLLEML-LURJTMIESA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
156.2 °F
Flash Point(C) 
69 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

69819 (−)-Butyl L-lactate, ≥97.0% (sum of enantiomers, GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?