• USA Home
  • 420190 - (R)-(+)-α-Acetoxyphenylacetonitrile

EMAIL THIS PAGE TO A FRIEND
420190 Sigma-Aldrich

(R)-(+)-α-Acetoxyphenylacetonitrile

99%

Synonym: (R)-α-Cyanobenzyl acetate, O-Acetyl-D-mandelonitrile

  • CAS Number 119718-89-7

  • Linear Formula C6H5CH(O2CCH3)CN

  • Molecular Weight 175.18

  •  Beilstein/REAXYS Number 4293296

  •  MDL number MFCD00191699

Purchase

Properties

assay   99%
optical activity   [α]20/D +8.0°, c = 10 in chloroform
refractive index   n20/D 1.506 (lit.)
bp   268 °C (lit.)
density   1.115 g/mL at 25 °C (lit.)
SMILES string   CC(=O)O[C@@H](C#N)c1ccccc1
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Precautionary statements 
RIDADR 
UN 3276 6.1 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
235.4 °F - closed cup
Flash Point(C) 
113 °C - closed cup
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?