• USA Home
  • 420387 - (1R)-(−)-Neomenthyl acetate

EMAIL THIS PAGE TO A FRIEND
420387 Aldrich

(1R)-(−)-Neomenthyl acetate

99%

Synonym: (−)-(1R,2R,5S)-Neomenthyl acetate

  • CAS Number 146502-80-9

  • Empirical Formula (Hill Notation) C12H22O2

  • Molecular Weight 198.30

  •  Beilstein Registry Number 1909420

  •  MDL number MFCD00062976

  •  PubChem Substance ID 24866334

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Esters, Organic Building Blocks More...
InChI Key   XHXUANMFYXWVNG-MVWJERBFSA-N
assay   99%
optical activity   [α]20/D −48°, c = 8 in benzene
bp   81 °C/3 mmHg(lit.)
mp   36-38 °C(lit.)
density   0.912 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1325 4.1 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?