• USA Home
  • 420514 - (1S,9S)-(−)-1,9-Bis[(tert-butyldimethylsilyloxy)methyl]-5-cyanosemicorrin

EMAIL THIS PAGE TO A FRIEND
420514 Sigma-Aldrich

(1S,9S)-(−)-1,9-Bis[(tert-butyldimethylsilyloxy)methyl]-5-cyanosemicorrin

97%

Synonym: (1S,9S)-1,9-Bis[(tert-butyldimethylsiloxy)methyl]-5-cyanosemicorrin, Semicorrin chiral ligand 2

  • CAS Number 105251-52-3

  • Empirical Formula (Hill Notation) C24H45N3O2Si2

  • Molecular Weight 463.80

  •  Beilstein/REAXYS Number 5153664

  •  MDL number MFCD00142698

  •  PubChem Substance ID 24866340

Purchase

Properties

assay   97%
optical activity   [α]20/D −64°, c = 1 in chloroform
mp   74-77 °C (lit.)

Description

Application

Ligand for enantioselective catalysis.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?