• USA Home
  • 421529 - Zonyl® FTS fluorotelomer intermediate

EMAIL THIS PAGE TO A FRIEND
421529 Aldrich

Zonyl® FTS fluorotelomer intermediate

average Mn ~703

Synonym: 2-(Perfluoroalkyl)ethyl stearate

  • Linear Formula C17H35CO2CH2CH2(CF2)nF, n~7-8

  •  MDL number MFCD00197963

  •  PubChem Substance ID 24866381

Purchase

Properties

Related Categories Fluorocarbon Surfactants, Materials Science, Micro/NanoElectronics, Self Assembly and Lithography, Self-Assembly Materials,
mol wt   average Mn ~703
composition   fluorine, 47 wt. %
viscosity   10 cP(60 °C)(lit.)
bp   348 °C(lit.)
mp   30-45 °C(lit.)
solubility   acetone, MEK and trichloroethylene: soluble
density   1.108 g/mL at 25 °C(lit.)

Description

Legal Information

DuPont Product

Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?