• USA Home
  • 421537 - Zonyl® TBC fluorotelomer intermediate

EMAIL THIS PAGE TO A FRIEND
421537 Aldrich

Zonyl® TBC fluorotelomer intermediate

average Mn ~1,563

Synonym: Triperfluoroalkyl citrate

  • CAS Number 71663-53-1

  • Linear Formula HOC[CO2CH2CH2(CF2)nF][CH2CO2CH2CH2(CF2)nF]2, n~8

  • Molecular Weight 1544.46

  •  MDL number MFCD00286561

  •  PubChem Substance ID 24866382

Purchase

Properties

Related Categories Fluorocarbon Surfactants, Materials Science, Micro/NanoElectronics, Self Assembly and Lithography, Self-Assembly Materials,
InChI Key   FORWMHUYULLNCN-UHFFFAOYSA-N
mol wt   average Mn ~1,563
composition   fluorine, 63 wt. %
viscosity   10 cP(70 °C)(lit.)
mp   50-70 °C (lit.)
density   1.5 g/mL at 25 °C(lit.)

Description

Legal Information

DuPont Product

Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?