• USA Home
  • 421677 - β-Asp-Phe methyl ester

EMAIL THIS PAGE TO A FRIEND
421677 Sigma-Aldrich

β-Asp-Phe methyl ester

96%

Synonym: N-β-L-Aspartyl-L-phenylalanine methyl ester

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Phenylalanine More...
Quality Level   100
assay   96%
optical activity   [α]20/D +6°, c = 1 in H2O
application(s)   peptide synthesis: suitable
impurities   <4% water
mp   201-204 °C (dec.) (lit.)
SMILES string   COC(=O)[C@H](Cc1ccccc1)NC(=O)C[C@H](N)C(O)=O
InChI   1S/C14H18N2O5/c1-21-14(20)11(7-9-5-3-2-4-6-9)16-12(17)8-10(15)13(18)19/h2-6,10-11H,7-8,15H2,1H3,(H,16,17)(H,18,19)/t10-,11-/m0/s1
InChI key   SHHIPKDJQYIJJF-QWRGUYRKSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?