• USA Home
  • 421766 - (S)-(+)-α-Methoxyphenylacetic acid

EMAIL THIS PAGE TO A FRIEND
421766 Sigma-Aldrich

(S)-(+)-α-Methoxyphenylacetic acid

≥99%, ChiraSelect

Synonym: O-Methyl-L-mandelic acid

  • CAS Number 26164-26-1

  • Linear Formula C6H5CH(OCH3)CO2H

  • Molecular Weight 166.17

  •  Beilstein/REAXYS Number 2361718

  •  EC Number 247-492-5

  •  MDL number MFCD00064216

Purchase

Properties

grade   ChiraSelect
assay   ≥99%
optical activity   [α]20/D +148°, c = 0.5 in ethanol
optical purity   ee: ≥99% (GLC)
mp   64-66 °C (lit.)
SMILES string   CO[C@H](C(O)=O)c1ccccc1
InChI   1S/C9H10O3/c1-12-8(9(10)11)7-5-3-2-4-6-7/h2-6,8H,1H3,(H,10,11)/t8-/m0/s1
InChI key   DIWVBIXQCNRCFE-QMMMGPOBSA-N

Description

Application

Reactant involved in the synthesis of biologically active molecules including:
• Acyclonucleoside phosphonates structural analogs of adefovir
• Labeled discodermolide for studying binding to tubulin
• 10-Isocyano-4-cadinene used for antifouling activity

Reactant involved in studies of immunostimulating chromanones gonytolides A-C

Reactant involved in:
• Hydroxylations and epoxidations
• Hydrogenations

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

248983 (S)-(+)-α-Methoxyphenylacetic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?