• USA Home
  • 422681 - trans-Perfluorodecalin

EMAIL THIS PAGE TO A FRIEND
422681 Aldrich

trans-Perfluorodecalin

98%

Synonym: trans-Octadecafluorodecahydronaphthalene

  • CAS Number 60433-12-7

  • Empirical Formula (Hill Notation) C10F18

  • Molecular Weight 462.08

  •  MDL number MFCD00153232

Purchase

Properties

assay   98%
refractive index   n20/D 1.313(lit.)
bp   140-142 °C(lit.)
mp   −175 °C(lit.)
density   1.92 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
UN 1993 3/PG 3
WGK Germany 
3
Flash Point(F) 
119 °F
Flash Point(C) 
48 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?