• USA Home
  • 422762 - (S)-(+)-2-Phenyl-N-(trifluoroacetyl)glycine

EMAIL THIS PAGE TO A FRIEND
422762 Aldrich

(S)-(+)-2-Phenyl-N-(trifluoroacetyl)glycine

99%

  • CAS Number 155894-96-5

  • Empirical Formula (Hill Notation) C10H8F3NO3

  • Molecular Weight 247.17

  •  MDL number MFCD00192397

Purchase

Properties

assay   99%
optical activity   [α]20/D +163°, c = 1 in methanol
mp   156-159 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?