• USA Home
  • 422800 - (S)-(−)-2-Acetoxysuccinic anhydride

EMAIL THIS PAGE TO A FRIEND
422800 Sigma-Aldrich

(S)-(−)-2-Acetoxysuccinic anhydride

98%

Synonym: (S)-3-Acetoxy-dihydro-2,5-furandione, O-Acetyl-L-malic anhydride

  • CAS Number 59025-03-5

  • Empirical Formula (Hill Notation) C6H6O5

  • Molecular Weight 158.11

  •  Beilstein Registry Number 82504

  •  MDL number MFCD00082708

  •  PubChem Substance ID 24866455

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acid Anhydrides, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   SSWJHSASZZAIAU-BYPYZUCNSA-N
assay   98%
optical activity   [α]20/D −27°, c = 1 in chloroform
optical purity   ee: 98% (GLC)
mp   54-56 °C (lit.)

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?