EMAIL THIS PAGE TO A FRIEND
423386 Aldrich

(S)-(+)-2-Octanol

99%, ChiraSelect

  • CAS Number 6169-06-8

  • Linear Formula CH3(CH2)5CH(OH)CH3

  • Molecular Weight 130.23

  •  Beilstein/REAXYS Number 1719323

  •  MDL number MFCD00064283

Purchase

Properties

grade   ChiraSelect
InChI Key   SJWFXCIHNDVPSH-QMMMGPOBSA-N
assay   99%
optical purity   ee: ≥99% (GLC)
refractive index   n20/D 1.426 (lit.)
bp   175 °C(lit.)
density   0.822 g/mL at 25 °C (lit.)

Description

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?