• USA Home
  • 424366 - Potassium peroxydiphosphate

EMAIL THIS PAGE TO A FRIEND
424366 Aldrich

Potassium peroxydiphosphate

technical grade

Purchase

Properties

grade   technical grade
impurities   15% K2HPO4
  5% KF

Description

Application

Initiator for the aqueous polymerization of methacrylamide and was also employed in a study of the oxidative polymerization of methylenebisacrylamide.

Safety & Documentation
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?