EMAIL THIS PAGE TO A FRIEND
425060 Sigma-Aldrich

Tetraallylsilane

97%

Synonym: Tetra(2-propenyl)silane

  • CAS Number 1112-66-9

  • Linear Formula (H2C=CHCH2)4Si

  • Molecular Weight 192.37

  •  Beilstein/REAXYS Number 1750943

  •  MDL number MFCD00048161

Purchase

Properties

InChI Key   AKRQMTFHUVDMIL-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.485 (lit.)
bp   90 °C/3 mmHg(lit.)
density   0.831 g/mL at 25 °C (lit.)

Description

Application

Basic building-block in dendrimer synthesis.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

86717 Tetraallylsilane, ≥97.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?