• USA Home
  • 425133 - 5-Fluoroorotic acid hydrate

EMAIL THIS PAGE TO A FRIEND
425133 Sigma-Aldrich

5-Fluoroorotic acid hydrate

98%

Synonym: 2,6-Dihydroxy-5-fluoropyrimidine-4-carboxylic acid, 5-Fluorouracil-4-carboxylic acid

Purchase

Properties

InChI Key   LODRRYMGPWQCTR-UHFFFAOYSA-N
assay   98%
mp   278 °C (dec.) (lit.)

Description

Application

Useful in the selection of orotidine-5′-phosphate decarboxylase mutants of Saccharomyces cerevisiae.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UV7800000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

F5013 5-Fluoroorotic acid hydrate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?