• USA Home
  • 425338 - Styryl 9M tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
425338 Aldrich

Styryl 9M tetrafluoroborate

Dye content 99 %

Synonym: 2-[[3-[2-[4-(Dimethylamino)phenyl]ethenyl]-5,5-dimethyl-2-cyclohexen-1-ylidene]methyl]-3-methylbenzothiazolium tetrafluoroborate

  • CAS Number 207683-25-8

  • Empirical Formula (Hill Notation) C27H31BF4N2S

  • Molecular Weight 502.42

Purchase

Properties

composition   Dye content, 99%
mp   240 °C (dec.) (lit.)
absorption   λmax 584 nm

Description

Application

Suitable as a laser dye

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?