• USA Home
  • 426474 - Trimethylamine solution

EMAIL THIS PAGE TO A FRIEND
426474 Sigma-Aldrich

Trimethylamine solution

25 wt. % in H2O

Purchase

Properties

Related Categories Amines, Building Blocks, C2 to C5, Chemical Synthesis, Nitrogen Compounds - Organic Building Blocks,
InChI Key   GETQZCLCWQTVFV-UHFFFAOYSA-N
concentration   25 wt. % in H2O
  25 wt. % in H2O
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1297 - class 3 - PG 2 - Trimethylamine, aqueous solutions
WGK Germany 
1
Flash Point(F) 
<3.2 °F
Flash Point(C) 
<-16 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?