• USA Home
  • 427268 - Poly(1,4-phenylene sulfide)

EMAIL THIS PAGE TO A FRIEND
427268 Sigma-Aldrich

Poly(1,4-phenylene sulfide)

pellets, viscosity 275 poise (310 °C)(lit.)

Purchase

Properties

Related Categories π-Conjugated Polymers, Materials Science, Poly(phenylene sulfide), Polymer Science, Polymers More...
form   pellets
hardness   93 (Rockwell M, ASTM D 785)
viscosity   275 poise(310 °C)(lit.)
mp   285-300 °C
density   1.35 g/mL at 25 °C

Description

Application

Auto intake manifolds and pumps, electrical connectors and circuit boards, flame-resistant clothing, valves, coatings, non-stick coatings, power tool parts and heater grills.

Packaging

100 g in poly bottle

Features and Benefits

High performance, linear thermoplastic. Excellent thermal and chemical resistance, inherent flame resistance and superior electrical insulation properties.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?