• USA Home
  • 429333 - Dysprosium(III) iodide

EMAIL THIS PAGE TO A FRIEND
429333 Aldrich

Dysprosium(III) iodide

anhydrous, flakes, 99.99% trace metals basis

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Dysprosium Catalysts, Dysprosium Salts, Essential Chemicals,
grade   anhydrous
InChI Key   RZQFCZYXPRKMTP-UHFFFAOYSA-K
assay   99.99% trace metals basis
form   flakes
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

The Rare Earth Crisis—The Supply/Demand Situation for 2010–2015

K.A. Gschneidner, Jr. Ames Laboratory, U.S. Department of Energy and Department of Materials Science and Engineering Iowa State University, Ames, Iowa 50011-3020, USA Email: cagey@ameslab.gov
Keywords: Absorption, Automotive, Capillary electrophoresis, Catalysis, Ceramics, Coloring agents, Detection methods, Environmental, Industries, Magnetic resonance imaging, Materials Science, Oxidations, Poisons, Reductions, Scintillation, Semiconductor, Separation, Substitutions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?