EMAIL THIS PAGE TO A FRIEND
429783 Sigma-Aldrich

2-Adamantanone

purified by sublimation, ≥99%

  • CAS Number 700-58-3

  • Empirical Formula (Hill Notation) C10H14O

  • Molecular Weight 150.22

  •  Beilstein/REAXYS Number 1210235

  •  EC Number 211-847-2

  •  MDL number MFCD00074737

  •  PubChem Substance ID 24866889

Purchase

Properties

assay   ≥99%
purified by   sublimation
mp   256-258 °C (subl.) (lit.)
SMILES string   O=C1C2CC3CC(C2)CC1C3
InChI   1S/C10H14O/c11-10-8-2-6-1-7(4-8)5-9(10)3-6/h6-9H,1-5H2/t6-,7+,8-,9+
InChI key   IYKFYARMMIESOX-SPJNRGJMSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AU5018000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?