EMAIL THIS PAGE TO A FRIEND
430366 Sigma-Aldrich

Antimony(V) sulfide

technical grade

Synonym: Antimony pentasulfide

Purchase

Properties

grade   technical grade
InChI Key   PPKVREKQVQREQD-UHFFFAOYSA-N
composition   Sb, 66%

Description

Other Notes

may contain a small amount of Sb2S3 and free sulfur

Safety & Documentation

Safety Information

RIDADR 
UN 3178 4.1/PG 2
WGK Germany 
2
RTECS 
CC6125000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

11133 Antimony(V) sulfide, technical, ≥60% Sb basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?