• USA Home
  • 432016 - 1,4-Dihydro-9-isopropylidene-1,4-methanonaphthalene

EMAIL THIS PAGE TO A FRIEND
432016 Aldrich

1,4-Dihydro-9-isopropylidene-1,4-methanonaphthalene

99%

  • CAS Number 7350-72-3

  • Empirical Formula (Hill Notation) C14H14

  • Molecular Weight 182.26

  •  MDL number MFCD00216595

Purchase

Properties

assay   99%
mp   90-93 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?