• USA Home
  • 434485 - Poly(methyl vinyl ether-alt-maleic anhydride), cross-linked with 1,9-decadiene

EMAIL THIS PAGE TO A FRIEND
434485 Sigma-Aldrich

Poly(methyl vinyl ether-alt-maleic anhydride), cross-linked with 1,9-decadiene

powder

Synonym: maleated copolymer, maleated polymer, maleic copolymer

Purchase

Properties

Related Categories Hydrophilic Polymers, Maleic Anhydride Copolymers, Materials Science, Miscellaneous Hydrophilic Polymers, Poly(methyl vinyl ether) and Copolymers,
InChI Key   UPBDXRPQPOWRKR-UHFFFAOYSA-N
form   powder
impurities   4.5%
viscosity   50,000-70,000 cP, 0.5 wt. % in H2O(25 °C, Brookfield)(lit.)

Description

Application

Component of hair and skin care products.

Features and Benefits

Forms gels at 0.25 to 0.5 wt. %. Produces non-tacky films.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?