• USA Home
  • 434949 - Polyacrylamide solution

EMAIL THIS PAGE TO A FRIEND
434949 Aldrich

Polyacrylamide solution

average Mw 10,000, 50 wt. % in H2O

Purchase

Properties

Related Categories Hydrophilic Polymers, Materials Science, Poly(N-isopropylacrylamide) (PNIPAM) and Polyacrylamide (PAM), Polyacrylamide (PAM), Polyacrylamide (PAM) and Copolymers,
mol wt   average Mw 10,000
concentration   50 wt. % in H2O
refractive index   n20/D 1.438
pH range   4 - 6
viscosity   5.5-9 cP, 15 % in H2O
density   1.189 g/mL at 25 °C

Description

Application

Binding agent, dispersing aid, lubricant, drag reduction and crystal formation control.

Features and Benefits

Tolerance for electrolytes, high initial tack and stable over a wide pH range.

Safety & Documentation

Safety Information

WGK Germany 
2
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Certificate of Origin


Webinar - Hydrogels
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?