• USA Home
  • 435538 - Ethylenediamine tetrakis(propoxylate-block-ethoxylate) tetrol

EMAIL THIS PAGE TO A FRIEND
435538 Sigma-Aldrich

Ethylenediamine tetrakis(propoxylate-block-ethoxylate) tetrol

average Mn ~15,000

Synonym: Tetronic 1107

Purchase

Properties

mol wt   average Mn ~15,000
surface tension   43 dyn/cm, 25 °C, 0.1 wt. % in H2O
viscosity   1,100(77 °C, Brookfield)(lit.)
mp   52 °C (DSC at peak)
transition temp   cloud point >100 °C (1 wt. % aqueous solution)
density   1.04 g/mL at 25 °C
HLB   24

Description

General description

Terminal secondary tetrahydroxy-functional oligomer.

Application

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Features and Benefits

Surface activity is dependent on the EO/PO ratio. Lower foaming, greater defoaming and reduced gelling tendencies relative to Pluronic® surfactants. Terminal secondary hydroxyl groups have lower reactivity and acidity than the primary hydroxyl groups in Pluronic® surfactants.

Legal Information

Pluronic is a registered trademark of BASF

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
1
Flash Point(F) 
>482 °F
Flash Point(C) 
>250 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

435511 Ethylenediamine tetrakis(propoxylate-block-ethoxylate) tetrol, average Mn ~3,600

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?