EMAIL THIS PAGE TO A FRIEND
435910 Sigma-Aldrich

Gallium antimonide

pieces, −6 mesh, ≥99.99%

Purchase

Properties

assay   ≥99.99%
form   pieces
particle size   −6 mesh
mp   980 °C (lit.)
density   5.62 g/mL at 25 °C (lit.)
SMILES string   [Ga]#[Sb]
InChI   1S/Ga.Sb
InChI key   VTGARNNDLOTBET-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?