• USA Home
  • 436119 - 2-Fluoro-3-(trifluoromethyl)phenol

EMAIL THIS PAGE TO A FRIEND
436119 Sigma-Aldrich

2-Fluoro-3-(trifluoromethyl)phenol

97%

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Fluorinated Building Blocks, Organic Building Blocks,
Quality Level   100
assay   97%
refractive index   n20/D 1.443 (lit.)
bp   122 °C (lit.)
density   1.431 g/mL at 25 °C (lit.)
SMILES string   Oc1cccc(c1F)C(F)(F)F
InChI   1S/C7H4F4O/c8-6-4(7(9,10)11)2-1-3-5(6)12/h1-3,12H
InChI key   DOICLFHNNMXINV-UHFFFAOYSA-N

Description

General description

2-Fluoro-3-(trifluoromethyl)phenol is an organic fluorinated building block.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Target organs 
Respiratory system
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
143.6 °F - closed cup
Flash Point(C) 
62 °C - closed cup

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?