• USA Home
  • 436453 - cis,cis-1,3,5-Tripropyl-1,3,5-cyclohexanetricarboxylic acid

EMAIL THIS PAGE TO A FRIEND
436453 Sigma-Aldrich

cis,cis-1,3,5-Tripropyl-1,3,5-cyclohexanetricarboxylic acid

97%

  • CAS Number 129063-52-1

  • Empirical Formula (Hill Notation) C18H30O6

  • Molecular Weight 342.43

  •  MDL number MFCD00274212

Purchase

Properties

assay   97%
mp   200-205 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?