• USA Home
  • 436577 - Poly(tetrahydrofuran) bis(3-aminopropyl) terminated

EMAIL THIS PAGE TO A FRIEND
436577 Aldrich

Poly(tetrahydrofuran) bis(3-aminopropyl) terminated

average Mn ~1,100

Purchase

Properties

Related Categories Ethers, Hydrophobic Polymers, Materials Science, Polymer Science, Polymers,
mol wt   average Mn ~1,100
refractive index   n20/D 1.465
bp   >200 °C(lit.)
mp   16-19 °C
density   0.98 g/mL at 25 °C
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN2 8 / PGII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?