• USA Home
  • 437050 - 2,3,4,6-Tetra-O-pivaloyl-β-D-galactopyranosyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
437050 Supelco

2,3,4,6-Tetra-O-pivaloyl-β-D-galactopyranosyl isothiocyanate

≥98%, ChiraSelect reagent

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology,
InChI Key   QCEYERYMOHEQHK-ICBNADEASA-N
assay   ≥98%
optical activity   [α]20/D +6.5°, c = 1 in chloroform
optical purity   ee: ≥99% (HPLC)
mp   142-145 °C (lit.)

Description

Packaging

100 mg in glass bottle

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

44891 2,3,4,6-Tetra-O-pivaloyl-β-D-glucopyranosyl isothiocyanate, for chiral derivatization, LiChropur, ≥95.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?