• USA Home
  • 437115 - (Trimethylsilyl)ketene bis(trimethylsilyl) acetal

EMAIL THIS PAGE TO A FRIEND
437115 Sigma-Aldrich

(Trimethylsilyl)ketene bis(trimethylsilyl) acetal

97%

  • CAS Number 65946-59-0

  • Linear Formula (CH3)3SiCH=C[OSi(CH3)3]2

  • Molecular Weight 276.60

  •  MDL number MFCD01632563

Purchase

Properties

assay   97%
refractive index   n20/D 1.428 (lit.)
bp   86 °C/10 mmHg (lit.)
density   0.848 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
105.8 °F
Flash Point(C) 
41 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?