• USA Home
  • 437131 - Trimethylsilyl (trimethylsilyl)acetate

EMAIL THIS PAGE TO A FRIEND
437131 Sigma-Aldrich

Trimethylsilyl (trimethylsilyl)acetate

97%

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   97%
refractive index   n20/D 1.417 (lit.)
bp   61 °C/10 mmHg (lit.)
density   0.867 g/mL at 25 °C (lit.)
SMILES string   C[Si](C)(C)CC(=O)O[Si](C)(C)C
InChI   1S/C8H20O2Si2/c1-11(2,3)7-8(9)10-12(4,5)6/h7H2,1-6H3
InChI key   LJHDMLCJRMZKGS-UHFFFAOYSA-N

Description

General description

Trimethylsilyl (trimethylsilyl)acetate is a trimethyl silyl ester.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?