• USA Home
  • 437182 - Ethyl 4-[[(ethylphenylamino)methylene]amino]benzoate

EMAIL THIS PAGE TO A FRIEND
437182 Sigma-Aldrich

Ethyl 4-[[(ethylphenylamino)methylene]amino]benzoate

99%

Purchase

Properties

assay   99%
bp   215 °C/2 mmHg(lit.)
mp   62-65 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?