• USA Home
  • 438553 - 4-Amino-5-phenyl-4H-1,2,4-triazole-3-thiol

EMAIL THIS PAGE TO A FRIEND
438553 Sigma-Aldrich

4-Amino-5-phenyl-4H-1,2,4-triazole-3-thiol

97%

  • CAS Number 22706-11-2

  • Empirical Formula (Hill Notation) C8H8N4S

  • Molecular Weight 192.24

  •  MDL number MFCD00269264

Purchase

Properties

InChI Key   OKNHZPGPLNUEPC-UHFFFAOYSA-N
assay   97%
mp   195 °C (dec.) (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?