• USA Home
  • 438634 - 6-Chloro-4-hydroxy-3-nitrocoumarin

EMAIL THIS PAGE TO A FRIEND
438634 Sigma-Aldrich

6-Chloro-4-hydroxy-3-nitrocoumarin

97%

  • CAS Number 24922-34-7

  • Empirical Formula (Hill Notation) C9H4ClNO5

  • Molecular Weight 241.58

  •  MDL number MFCD00674008

Purchase

Properties

assay   97%
SMILES string   OC1=C(C(=O)Oc2ccc(Cl)cc12)[N+]([O-])=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?