• USA Home
  • 441570 - (R)-(+)-2-Acetoxysuccinic anhydride

EMAIL THIS PAGE TO A FRIEND
441570 Aldrich

(R)-(+)-2-Acetoxysuccinic anhydride

96%

Synonym: (R)-3-Acetoxy-dihydro-2,5-furandione, O-Acetyl-D-malic anhydride

  • CAS Number 79814-40-7

  • Empirical Formula (Hill Notation) C6H6O5

  • Molecular Weight 158.11

  •  Beilstein Registry Number 130824

  •  MDL number MFCD00145214

  •  PubChem Substance ID 24867669

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acid Anhydrides, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   SSWJHSASZZAIAU-SCSAIBSYSA-N
assay   96%
optical activity   [α]20/D +27°, c = 1 in chloroform
optical purity   ee: 99% (GLC)
mp   56-58 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?