• USA Home
  • 441708 - 1-[[3-Ethyl-2(3H)-benzothiazolylidene]ethylidene]-2(1H)-naphthalenone

EMAIL THIS PAGE TO A FRIEND
441708 Sigma-Aldrich

1-[[3-Ethyl-2(3H)-benzothiazolylidene]ethylidene]-2(1H)-naphthalenone

Dye content 90 %

Purchase

Properties

Related Categories Cell Biology, E-F, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   JZFXWYPVXGMIKL-CVPKZDCNSA-N
composition   Dye content, 90%
mp   181-186 °C (lit.)
λmax   574 nm
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?