• USA Home
  • 442127 - Hydrogen tetrabromoaurate(III) hydrate

EMAIL THIS PAGE TO A FRIEND
442127 Sigma-Aldrich

Hydrogen tetrabromoaurate(III) hydrate

Synonym: Bromoauric acid

Purchase

Properties

Related Categories Essential Chemicals, Gold Salts, Materials Science, Metal and Ceramic Science, Research Essentials,
form   crystalline
SMILES string   O.Br.Br[Au](Br)Br
InChI   1S/Au.4BrH.H2O/h;4*1H;1H2/q+3;;;;;/p-3
InChI key   CGZJYXDZPHUJON-UHFFFAOYSA-K
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3260 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?