EMAIL THIS PAGE TO A FRIEND
442208 Sigma-Aldrich

Methyl perillate

97%

  • CAS Number 26460-67-3

  • Empirical Formula (Hill Notation) C11H16O2

  • Molecular Weight 180.24

Purchase

Properties

assay   97%
refractive index   n20/D 1.491 (lit.)
bp   68-72 °C/0.04 mmHg (lit.)
density   0.999 g/mL at 25 °C (lit.)
SMILES string   COC(=O)C1=CC[C@H](CC1)C(C)=C
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?