• USA Home
  • 442267 - 4,4′-Isopropylidenebis(diisodecyl phenyl phosphite)

EMAIL THIS PAGE TO A FRIEND
442267 Sigma-Aldrich

4,4′-Isopropylidenebis(diisodecyl phenyl phosphite)

  • CAS Number 61670-79-9

  • Linear Formula [(C10H21O)2POC6H4]2C(CH3)2

  • Molecular Weight 917.31

  •  MDL number MFCD00274626

Purchase

Properties

refractive index   n20/D 1.498 (lit.)
bp   336 °C (lit.)
density   0.964 g/mL at 25 °C (lit.)
SMILES string   CC(C)CCCCCCCOP(OCCCCCCCC(C)C)Oc1ccc(cc1)C(C)(C)c2ccc(OP(OCCCCCCCC(C)C)OCCCCCCCC(C)C)cc2

Description

Application

Low volatility heat stabilizer for PVC

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?