• USA Home
  • 442771 - 8-Quinolinyl trifluoromethanesulfonate

EMAIL THIS PAGE TO A FRIEND
442771 Sigma-Aldrich

8-Quinolinyl trifluoromethanesulfonate

97%

Synonym: 8-Quinolinyl triflate

Purchase

Properties

Related Categories Building Blocks, C8 to C10, Chemical Synthesis, Heterocyclic Building Blocks, Quinolines More...
InChI Key   YUWOECMFUBVGSE-UHFFFAOYSA-N
assay   97%
mp   65-68 °C (lit.)

Description

Application

Employed in the palladium-catalyzed β-arylation of modified vinyl ethers.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?